Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005990.hg.1 |
ENST00000278888.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005990.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005990.hg.1 |
TC11000505.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005988.hg.1 |
ENST00000278888.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005988.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005988.hg.1 |
TC11000505.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005989.hg.1 |
ENST00000278888.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005989.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005989.hg.1 |
TC11000505.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000524868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000524868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000524868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154674.hg.1 |
TC1100007765.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
ENST00000524868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
ENST00000524868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154677.hg.1 |
TC1100007765.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000524868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000524868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000524868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007765.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
ENST00000524868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
ENST00000278888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
ENST00000440896.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
ENST00000524868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
ENST00000617306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
NM_000139.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
NM_001256916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100154678.hg.1 |
TC1100007765.hg.1 |
Clariom D TC |