Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000493174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000493174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000493174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000493174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
ENST00000493174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022701.hg.1 |
TC07001507.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022701.hg.1 |
ENST00000297873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022701.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000458679.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000493174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000493174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000493174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000493174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
ENST00000493174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011504.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181799.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181799.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181799.hg.1 |
ENST00000297873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181799.hg.1 |
NM_152559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181799.hg.1 |
TC0700011504.hg.1 |
Clariom D TC |