Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017084.hg.1 |
ENST00000254951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017084.hg.1 |
ENST00000338779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017084.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017084.hg.1 |
TC16001237.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000562883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000562883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000562883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000562883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000562883.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017082.hg.1 |
ENST00000254951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017082.hg.1 |
ENST00000338779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017082.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017082.hg.1 |
TC16001237.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000562883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000562883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000562883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000562883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000562883.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011560.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194278.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194278.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194278.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194278.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194278.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194278.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194278.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194278.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194278.hg.1 |
TC1600011560.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600194274.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194274.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194274.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194274.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194274.hg.1 |
ENST00000338779.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194274.hg.1 |
ENST00000576338.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194274.hg.1 |
ENST00000616026.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194274.hg.1 |
NM_138383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194274.hg.1 |
TC1600011560.hg.1 |
Clariom D TC |