Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005256064.4 | XP_005256121.1 | 1 - 2 | 198 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |