Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286474.1 | NP_001273403.1 | 9 - 10 | 488 | 108 | |
NM_001286475.1 | NP_001273404.1 | 8 - 9 | 467 | 108 | ||
GenBank mRNA | AK225742.1 | - | 9 - 10 | 479 | 108 | |
AK302176.1 | - | 8 - 9 | 427 | 108 | ||
AK310230.1 | - | 8 - 9 | 456 | 108 | ||
U60665.1 | - | 9 - 10 | 518 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |