Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020703.hg.1 |
TC06001732.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020703.hg.1 |
ENST00000340840.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020703.hg.1 |
ENST00000354325.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020703.hg.1 |
ENST00000372917.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020703.hg.1 |
ENST00000372922.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020703.hg.1 |
ENST00000541110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020703.hg.1 |
NM_033502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
ENST00000340840.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
ENST00000354325.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
ENST00000372917.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
ENST00000372922.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
ENST00000541110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
NM_001297573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
NM_033502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
ENST00000340840.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
ENST00000354325.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
ENST00000372917.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
ENST00000372922.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
ENST00000541110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
NM_001297573.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
NM_033502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181229.hg.1 |
TC0600011823.hg.1 |
Clariom D TC |