Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000388887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000548073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000548073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000548073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000548073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000548073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000548579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000548579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000548579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000548579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000548579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000549474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000549474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000549474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000549474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000549798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000549798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000549798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000552777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000552777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000552777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
ENST00000552777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008659.hg.1 |
NM_017564.9 |
Refseq |