Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000573377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000573377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000573377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
ENST00000573377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007422.hg.1 |
TC17002878.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007422.hg.1 |
ENST00000262017.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007422.hg.1 |
ENST00000559488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007422.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202755.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202755.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202755.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202755.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202755.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202755.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202755.hg.1 |
TC1700012274.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000559488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000571680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000573377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000573377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000573377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
ENST00000573377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012274.hg.1 |
NM_000212.2 |
Refseq |