Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000459581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008473.hg.1 |
TC09000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008473.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008473.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008472.hg.1 |
TC09000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008472.hg.1 |
ENST00000309971.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008472.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008472.hg.1 |
ENST00000539582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008472.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008472.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008471.hg.1 |
TC09000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008471.hg.1 |
ENST00000309971.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008471.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008471.hg.1 |
ENST00000539582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008471.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008471.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184794.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184794.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184794.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184794.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184794.hg.1 |
TC0900012178.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900184792.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184792.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184792.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184792.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184792.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184792.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184792.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184792.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184792.hg.1 |
TC0900012178.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900184790.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184790.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184790.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184790.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184790.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184790.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184790.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184790.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184790.hg.1 |
TC0900012178.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000309971.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000372770.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000459581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
ENST00000494417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001003722.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012178.hg.1 |
NM_001499.2 |
Refseq |