Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000519806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000522623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000522623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000522623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000522623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000522623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
ENST00000523823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014962.hg.1 |
TC08001164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014962.hg.1 |
ENST00000297737.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014962.hg.1 |
ENST00000315769.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014962.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014962.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014961.hg.1 |
TC08001164.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014961.hg.1 |
ENST00000297737.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014961.hg.1 |
ENST00000315769.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014961.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08014961.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000519806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000522623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000522623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000522623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000522623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000522623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
ENST00000523823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010224.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000519806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000519806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169575.hg.1 |
TC0800010224.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000297737.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000315769.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000518242.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000519406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000523188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
ENST00000523542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
NM_001135731.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
NM_024645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800169576.hg.1 |
TC0800010224.hg.1 |
Clariom D TC |