Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028417.hg.1 |
TC06002263.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028417.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028417.hg.1 |
ENST00000367089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028417.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367085.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367085.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367085.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367085.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194593.hg.1 |
TC0600013719.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367085.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367085.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367085.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367085.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013719.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
ENST00000367088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
ENST00000367089.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
NM_001291602.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
NM_001291603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
NM_006519.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194594.hg.1 |
TC0600013719.hg.1 |
Clariom D TC |