Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000371347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000411689.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603486.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000603501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604127.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
ENST00000604908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204051.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_001204052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008166.hg.1 |
NM_004277.4 |
Refseq |