Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000572234.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000572234.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000572234.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
ENST00000575868.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009100.hg.1 |
ENST00000226067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009100.hg.1 |
ENST00000430986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009100.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009100.hg.1 |
TC17000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009095.hg.1 |
ENST00000226067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009095.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009095.hg.1 |
TC17000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009097.hg.1 |
ENST00000226067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009097.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009097.hg.1 |
TC17000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009094.hg.1 |
ENST00000226067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009094.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009094.hg.1 |
TC17000693.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700162105.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162105.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162105.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162105.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162105.hg.1 |
TC1700008331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162111.hg.1 |
TC1700008331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700162102.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162102.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162102.hg.1 |
TC1700008331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162110.hg.1 |
TC1700008331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700162104.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162104.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162104.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162104.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162104.hg.1 |
TC1700008331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000226067.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000430986.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000570962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000572002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000572234.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000572234.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000572234.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000573945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
ENST00000575868.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008331.hg.1 |
NM_002126.4 |
Refseq |