Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007243.hg.1 |
ENST00000242848.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007243.hg.1 |
ENST00000282007.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007243.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007243.hg.1 |
TC13000640.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000428921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000428921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000428921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000428921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000428921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000464597.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000464597.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000464597.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000464597.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000470308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000470308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000470308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
ENST00000470308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007241.hg.1 |
ENST00000242848.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007241.hg.1 |
ENST00000282007.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007241.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007241.hg.1 |
TC13000640.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007244.hg.1 |
ENST00000242848.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007244.hg.1 |
ENST00000282007.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007244.hg.1 |
ENST00000431251.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007244.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13007244.hg.1 |
TC13000640.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000428921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000428921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000428921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000428921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000428921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000464597.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000464597.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000464597.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000464597.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000470308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000470308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000470308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
ENST00000470308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008837.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159600.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159600.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159600.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159600.hg.1 |
ENST00000242848.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159600.hg.1 |
ENST00000282007.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300159600.hg.1 |
NM_015070.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300159600.hg.1 |
TC1300008837.hg.1 |
Clariom D TC |