Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
TC05001366.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
ENST00000336909.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
ENST00000381287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
ENST00000381294.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
ENST00000381298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
ENST00000502326.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
ENST00000522633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
ENST00000536319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
NM_001190981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
NM_002184.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019128.hg.1 |
NM_175767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000336909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000381287.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000381294.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000381298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000502326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000503773.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000506241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000522633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
NM_001190981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
NM_002184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
NM_175767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
NR_120480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000336909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000381287.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000381294.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000381298.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000502326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000503773.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000506241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
ENST00000522633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
NM_001190981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
NM_002184.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
NM_175767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
NR_120480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175290.hg.1 |
TC0500010788.hg.1 |
Clariom D TC |