Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159767.1 | NP_001153239.1 | 1 - 2 | 174 | 60 | |
NR_027624.1 | 1 - 2 | 174 | 60 | |||
GenBank mRNA | AK297496.1 | - | 1 - 2 | 165 | 60 | |
BC107757.1 | - | 1 - 2 | 132 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |