Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126336.2 | NP_001119808.1 | 6 - 7 | 1400 | 70 | |
GenBank mRNA | D32039.1 | - | 6 - 7 | 1149 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Inventoried Probe spans exons |