Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
ENST00000433100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
ENST00000434449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
ENST00000525518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
ENST00000526609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
ENST00000527179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
ENST00000530046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
ENST00000531459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
NM_000694.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
NM_001030010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
NM_001161473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009104.hg.1 |
TC11000702.hg.1 |
HTA TC |