Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001692.hg.1 |
TC09000156.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001692.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001691.hg.1 |
TC09000156.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001691.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001695.hg.1 |
TC09000156.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001695.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001695.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001694.hg.1 |
TC09000156.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001694.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001694.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147536.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147536.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147536.hg.1 |
TC0900007010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900147538.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147538.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147538.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147538.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147538.hg.1 |
TC0900007010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900147539.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147539.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147539.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147539.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147539.hg.1 |
TC0900007010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900147535.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147535.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147535.hg.1 |
TC0900007010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900147541.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147541.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147541.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900147541.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900147541.hg.1 |
TC0900007010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
ENST00000263228.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007010.hg.1 |
NM_017811.3 |
Refseq |