Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013854.hg.1 |
ENST00000264033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013854.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013854.hg.1 |
TC11001089.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013855.hg.1 |
ENST00000264033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013855.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013855.hg.1 |
TC11001089.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013856.hg.1 |
ENST00000264033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013856.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11013856.hg.1 |
TC11001089.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100169332.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169332.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169332.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169332.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169332.hg.1 |
TC1100009245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100169330.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169330.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169330.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169330.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169330.hg.1 |
TC1100009245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100169331.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169331.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169331.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169331.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169331.hg.1 |
TC1100009245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100169333.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169333.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169333.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100169333.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100169333.hg.1 |
TC1100009245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
ENST00000264033.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009245.hg.1 |
NM_005188.3 |
Refseq |