Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010144.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010144.hg.1 |
TC19000730.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010149.hg.1 |
ENST00000221498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010149.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010149.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010149.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010149.hg.1 |
TC19000730.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900165861.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165861.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165861.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165861.hg.1 |
TC1900008535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165861.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165861.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165861.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000594268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
ENST00000598682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008535.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
NM_014419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
TC1900008535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
ENST00000221498.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
ENST00000596402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
ENST00000597546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
ENST00000597873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
NM_001197301.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
NM_001197302.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165869.hg.1 |
NM_014419.3 |
Refseq |