Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036378.hg.1 |
TC02002263.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036378.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036378.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036377.hg.1 |
TC02002263.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036377.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036377.hg.1 |
ENST00000537910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036377.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203039.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203039.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203039.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203039.hg.1 |
ENST00000263707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203039.hg.1 |
ENST00000464621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203039.hg.1 |
NM_014553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203039.hg.1 |
TC0200014085.hg.1 |
Clariom D TC |