Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000475778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000475778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000475778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
ENST00000475778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001388.hg.1 |
TC0X000102.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001388.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001388.hg.1 |
ENST00000418858.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001388.hg.1 |
ENST00000535894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001388.hg.1 |
ENST00000537599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001388.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000475778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000475778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000475778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
ENST00000475778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006775.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146398.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146398.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146398.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146398.hg.1 |
ENST00000379374.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146398.hg.1 |
NM_000444.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146398.hg.1 |
NM_001282754.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146398.hg.1 |
TC0X00006775.hg.1 |
Clariom D TC |