Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006995.hg.1 |
TC0X000523.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006995.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006995.hg.1 |
ENST00000372453.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006995.hg.1 |
ENST00000535523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006995.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006995.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155138.hg.1 |
TC0X00008064.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000336387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000372453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
ENST00000535523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_001169154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008064.hg.1 |
NM_173494.1 |
Refseq |