Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020440.hg.1 |
ENST00000302503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020440.hg.1 |
ENST00000534074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020440.hg.1 |
ENST00000542981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020440.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020440.hg.1 |
TC11001746.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181573.hg.1 |
TC1100010757.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000302503.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000525649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000530428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000533571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000534074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
ENST00000539759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010757.hg.1 |
NM_014342.3 |
Refseq |