Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316341.1 | NP_001303270.1 | 7 - 8 | 903 | 126 | |
NM_002816.4 | NP_002807.1 | 5 - 6 | 619 | 126 | ||
NM_174871.3 | NP_777360.1 | 4 - 5 | 559 | 126 | ||
GenBank mRNA | AB003103.1 | - | 5 - 6 | 554 | 126 | |
AK091198.1 | - | 4 - 5 | 537 | 126 | ||
AK222679.1 | - | 5 - 6 | 536 | 126 | ||
AK307821.1 | - | 5 - 6 | 569 | 126 | ||
AK308883.1 | - | 5 - 6 | 560 | 126 | ||
BC019062.2 | - | 5 - 6 | 550 | 4111477 | 126 | |
BC065826.1 | - | 7 - 8 | 823 | 30386487 | 126 | |
KU178288.1 | - | 5 - 6 | 511 | 126 | ||
KU178289.1 | - | 5 - 6 | 511 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |