Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011355.hg.1 |
TC03000630.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011355.hg.1 |
ENST00000264474.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011355.hg.1 |
ENST00000479204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011355.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011354.hg.1 |
TC03000630.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011354.hg.1 |
ENST00000264474.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011354.hg.1 |
ENST00000479204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011354.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011351.hg.1 |
TC03000630.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03011351.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160843.hg.1 |
ENST00000264474.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160843.hg.1 |
ENST00000479204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160843.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160843.hg.1 |
ENST00000264474.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160843.hg.1 |
ENST00000479204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160843.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160843.hg.1 |
TC0300008550.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300160842.hg.1 |
ENST00000264474.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160842.hg.1 |
ENST00000479204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160842.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160842.hg.1 |
ENST00000264474.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160842.hg.1 |
ENST00000479204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160842.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160842.hg.1 |
TC0300008550.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300160839.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160839.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160839.hg.1 |
TC0300008550.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300160844.hg.1 |
ENST00000264474.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160844.hg.1 |
ENST00000479204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160844.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160844.hg.1 |
ENST00000264474.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160844.hg.1 |
ENST00000479204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160844.hg.1 |
NM_005213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160844.hg.1 |
TC0300008550.hg.1 |
Clariom D TC |