Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000578159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000578159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
ENST00000584496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024950.hg.1 |
ENST00000293190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024950.hg.1 |
ENST00000347612.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024950.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024950.hg.1 |
TC17001863.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000578159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000578159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
ENST00000584496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011719.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
ENST00000293190.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
ENST00000347612.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
ENST00000584176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
NM_000835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
NM_001278553.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
NR_103735.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194883.hg.1 |
TC1700011719.hg.1 |
Clariom D TC |