Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029960.hg.1 |
TC02001878.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029960.hg.1 |
ENST00000233741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029960.hg.1 |
ENST00000402135.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029960.hg.1 |
ENST00000403295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029960.hg.1 |
ENST00000403676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029960.hg.1 |
NM_001114636.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029960.hg.1 |
NM_018062.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000233741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000402135.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000403295.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000403676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000427708.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000446381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000449070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000470506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
NM_001114636.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
NM_018062.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000233741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000402135.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000403295.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000403676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000427708.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000446381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000449070.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
ENST00000470506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
NM_001114636.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
NM_018062.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192520.hg.1 |
TC0200012684.hg.1 |
Clariom D TC |