Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018072.hg.1 |
TC03001020.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018072.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018072.hg.1 |
ENST00000411641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018072.hg.1 |
ENST00000541510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018072.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018074.hg.1 |
TC03001020.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018074.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018074.hg.1 |
ENST00000411641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018074.hg.1 |
ENST00000541510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03018074.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171058.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171058.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171058.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171058.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171058.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171058.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171058.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171058.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171058.hg.1 |
TC0300009772.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300171056.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171056.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171056.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171056.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171056.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171056.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171056.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171056.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171056.hg.1 |
TC0300009772.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000273784.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000411641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
ENST00000478441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009772.hg.1 |
NM_001622.3 |
Refseq |