Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012049.hg.1 |
TC07003341.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012049.hg.1 |
ENST00000000233.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012049.hg.1 |
ENST00000467281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012049.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000459680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000459680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000459680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000459680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000459680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000459680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198796.hg.1 |
TC0700013456.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000000233.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000415666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000459680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000459680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000459680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000459680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000463733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000467281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
ENST00000489673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013456.hg.1 |
NM_001662.3 |
Refseq |