Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011075.hg.1 |
TC0X000807.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011075.hg.1 |
ENST00000217939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011075.hg.1 |
ENST00000381114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011075.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011076.hg.1 |
TC0X000807.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011076.hg.1 |
ENST00000217939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011076.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011077.hg.1 |
TC0X000807.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011077.hg.1 |
ENST00000217939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011077.hg.1 |
ENST00000381114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011077.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011074.hg.1 |
TC0X000807.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011074.hg.1 |
ENST00000217939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011074.hg.1 |
ENST00000381114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011074.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161449.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161449.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161449.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161449.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161449.hg.1 |
TC0X00008940.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008940.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161446.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161446.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161446.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161446.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161446.hg.1 |
TC0X00008940.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006619.hg.1 |
ENST00000420610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161447.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161447.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161447.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161447.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161447.hg.1 |
TC0X00008940.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00161448.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161448.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161448.hg.1 |
ENST00000217939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00161448.hg.1 |
NM_015419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00161448.hg.1 |
TC0X00008940.hg.1 |
Clariom D TC |