Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015715.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015715.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015715.hg.1 |
TC12001186.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015716.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015716.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015716.hg.1 |
TC12001186.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200174047.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174047.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174047.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174047.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174047.hg.1 |
TC1200009846.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200174050.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174050.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174050.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174050.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174050.hg.1 |
TC1200009846.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200174052.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174052.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174052.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174052.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174052.hg.1 |
TC1200009846.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200174048.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174048.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174048.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174048.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174048.hg.1 |
TC1200009846.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174049.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174049.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174049.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174049.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174049.hg.1 |
TC1200009846.hg.1 |
Clariom D TC |