Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002798.hg.1 |
TC6_ssto_hap7000142.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002798.hg.1 |
NM_025258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003130.hg.1 |
TC6_cox_hap2000170.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2003130.hg.1 |
NM_025258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017825.hg.1 |
TC06001542.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017825.hg.1 |
ENST00000375686.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017825.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017825.hg.1 |
ENST00000447450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017825.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017825.hg.1 |
NM_025258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002797.hg.1 |
TC6_mcf_hap5000147.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002797.hg.1 |
NM_025258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003037.hg.1 |
TC6_dbb_hap3000159.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3003037.hg.1 |
NM_025258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017822.hg.1 |
TC06001542.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017822.hg.1 |
ENST00000375686.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017822.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017822.hg.1 |
ENST00000447450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017822.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017822.hg.1 |
NM_025258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001188.hg.1 |
TC6_apd_hap1000084.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001188.hg.1 |
NM_025258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000487013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000487013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000497645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000497645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000497645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011454.hg.1 |
ENST00000497645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000375688.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000467576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000486423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000487013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000487013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000497645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000497645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000497645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011454.hg.1 |
ENST00000497645.1 |
Ensembl |