Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000403050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000423760.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000469407.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000469407.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000469407.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000469407.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000470665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000470665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000470665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000470665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000470665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000470665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000473050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000473050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000473050.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000476009.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000476009.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000476009.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000476009.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000476009.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000476009.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000476009.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000476774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000476774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000481418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000481418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000481418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000481418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000481418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000481418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000481418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000489305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000489305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000489305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000490957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000493922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000493922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000493922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000493922.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000497851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000497851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000497851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000497851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000498784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000498784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000498784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000498784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
ENST00000521747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NM_014660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006715.hg.1 |
NR_033436.1 |
Refseq |