Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000347.5 | NP_000338.3 | 17 - 18 | 3908 | 67 | |
NM_001024858.2 | NP_001020029.1 | 17 - 18 | 3908 | 67 | ||
XM_005268023.4 | XP_005268080.1 | 18 - 19 | 3975 | 67 | ||
XM_011537105.2 | XP_011535407.1 | 18 - 19 | 3975 | 67 | ||
XM_017021612.1 | XP_016877101.1 | 19 - 20 | 4123 | 67 | ||
XM_017021613.1 | XP_016877102.1 | 18 - 19 | 3975 | 67 | ||
XM_017021614.1 | XP_016877103.1 | 18 - 19 | 3975 | 67 | ||
GenBank mRNA | AB209415.1 | - | 18 - 19 | 3997 | 67 | |
BC136284.1 | - | 17 - 18 | 3887 | 9020271 | 67 | |
BC136285.1 | - | 17 - 18 | 3887 | 9020272 | 67 | |
J05500.1 | - | 18 - 19 | 3949 | 67 | ||
M18054.1 | - | 2 - 3 | 230 | 67 | ||
M37884.1 | - | 3 - 4 | 692 | 67 | ||
M57948.1 | - | 4 - 5 | 1075 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |