Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026585.hg.1 |
ENST00000334289.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026585.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026585.hg.1 |
TC11002223.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
ENST00000534527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012105.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193067.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193067.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193067.hg.1 |
ENST00000334289.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193067.hg.1 |
NM_178127.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193067.hg.1 |
TC1100012105.hg.1 |
Clariom D TC |