Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012097.hg.1 |
TC03000672.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012097.hg.1 |
ENST00000254730.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012097.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012097.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012098.hg.1 |
TC03000672.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012098.hg.1 |
ENST00000254730.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012098.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161990.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161990.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161990.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161990.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161990.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161990.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161990.hg.1 |
TC0300008715.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300161992.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161992.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161992.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161992.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161992.hg.1 |
TC0300008715.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000254730.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000483569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
ENST00000484438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008715.hg.1 |
NM_021937.4 |
Refseq |