Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018765.hg.1 |
TC02001224.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018765.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02018765.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010586.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200175006.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200175006.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200175006.hg.1 |
ENST00000272852.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200175006.hg.1 |
NM_173077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200175006.hg.1 |
TC0200010586.hg.1 |
Clariom D TC |