Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005629.hg.1 |
ENST00000170564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005629.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005629.hg.1 |
TC19000432.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000590062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000590062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000590062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000590062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157026.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157026.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157026.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157026.hg.1 |
TC1900007763.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900157026.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157026.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157026.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000170564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000590062.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000590062.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000590062.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000590062.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
ENST00000592262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007763.hg.1 |
NM_018025.2 |
Refseq |