Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02017470.hg.1 |
TC02005007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02017470.hg.1 |
ENST00000295079.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02017470.hg.1 |
ENST00000392290.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02017470.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000435773.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000435773.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000435773.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000435773.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000435773.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000469156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000469156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000469156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
ENST00000469156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000435773.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000435773.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000435773.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000435773.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000435773.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000469156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000469156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000469156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
ENST00000469156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016590.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200224663.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200224663.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200224663.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200224663.hg.1 |
ENST00000295079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200224663.hg.1 |
ENST00000392290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200224663.hg.1 |
NM_024520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200224663.hg.1 |
TC0200016590.hg.1 |
Clariom D TC |