Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002178.hg.1 |
TC6_dbb_hap3000120.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002178.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016789.hg.1 |
TC06001495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016789.hg.1 |
ENST00000376728.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016789.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016789.hg.1 |
ENST00000496062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016789.hg.1 |
ENST00000540829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016789.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002248.hg.1 |
TC6_cox_hap2000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002248.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001969.hg.1 |
TC6_mcf_hap5000111.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001969.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001972.hg.1 |
TC6_mcf_hap5000111.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001972.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002181.hg.1 |
TC6_dbb_hap3000120.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002181.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002223.hg.1 |
TC6_qbl_hap6000121.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002223.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001771.hg.1 |
TC6_mann_hap4000109.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001771.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002226.hg.1 |
TC6_qbl_hap6000121.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002226.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002022.hg.1 |
TC6_ssto_hap7000112.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002022.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002025.hg.1 |
TC6_ssto_hap7000112.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002025.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002251.hg.1 |
TC6_cox_hap2000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002251.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001768.hg.1 |
TC6_mann_hap4000109.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001768.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016792.hg.1 |
TC06001495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016792.hg.1 |
ENST00000376728.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016792.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016792.hg.1 |
ENST00000496062.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016792.hg.1 |
ENST00000540829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016792.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000480808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000480808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000480808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000484583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000484583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000484583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000484583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000484583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000493404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000493404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000493404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
ENST00000493404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000471695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000480808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000480808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000480808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000484583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000484583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000484583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000484583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000484583.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000493404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000493404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000493404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
ENST00000493404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011351.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600176419.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176419.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176419.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176419.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600176419.hg.1 |
ENST00000376734.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176419.hg.1 |
ENST00000468264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176419.hg.1 |
ENST00000485864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176419.hg.1 |
NM_007028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600176419.hg.1 |
TC0600011351.hg.1 |
Clariom D TC |