Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000355076.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000404589.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000404589.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000404589.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000404589.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000404589.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000404589.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000404589.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000404589.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000479179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000479179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000479179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000479179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000479470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000479470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000479470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000479470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000479470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000490264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000490264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000490264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000490264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000490264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000490264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000490264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000490264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000490264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000491093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000491093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000491093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000491093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000491093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
ENST00000491093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_001128214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_001128214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_001128214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_001128214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_001128214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_001128214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_001128214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_001128214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_153331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_153331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_153331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_153331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_153331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_153331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_153331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_153331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_153331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007617.hg.1 |
NM_153331.3 |
Refseq |