Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010160.hg.1 |
ENST00000481845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037118.hg.1 |
TC03002091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037118.hg.1 |
ENST00000296277.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037118.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037118.hg.1 |
ENST00000455270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037118.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037119.hg.1 |
TC03002091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037119.hg.1 |
ENST00000296277.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037119.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037119.hg.1 |
ENST00000455270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037119.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010478.hg.1 |
ENST00000453523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013458.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013458.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200529.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200529.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200529.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200529.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200529.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200529.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200529.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200529.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200529.hg.1 |
TC0300013458.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010478.hg.1 |
ENST00000453523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200528.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200528.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200528.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200528.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200528.hg.1 |
ENST00000296277.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200528.hg.1 |
ENST00000433055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200528.hg.1 |
ENST00000455270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200528.hg.1 |
NM_052969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200528.hg.1 |
TC0300013458.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010160.hg.1 |
ENST00000481845.1 |
Ensembl |