Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015215.hg.1 |
ENST00000320591.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015215.hg.1 |
ENST00000439553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015215.hg.1 |
ENST00000534837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015215.hg.1 |
ENST00000540656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015215.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015215.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015215.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015215.hg.1 |
TC12001148.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000536467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000536467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000542740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000542740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000542740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000542740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015214.hg.1 |
ENST00000320591.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015214.hg.1 |
ENST00000439553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015214.hg.1 |
ENST00000534837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015214.hg.1 |
ENST00000540656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015214.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015214.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015214.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015214.hg.1 |
TC12001148.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173080.hg.1 |
TC1200009749.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173078.hg.1 |
TC1200009749.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000320591.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000534837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000536467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000536467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000540656.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000542740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000542740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000542740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000542740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
ENST00000545917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_001244015.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009749.hg.1 |
NM_153685.3 |
Refseq |