Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012595.hg.1 |
ENST00000301295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012595.hg.1 |
ENST00000346986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012595.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19012595.hg.1 |
TC19000913.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008934.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170135.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170135.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170135.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170135.hg.1 |
NM_134444.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900170135.hg.1 |
TC1900008934.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900170135.hg.1 |
ENST00000301295.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170135.hg.1 |
ENST00000587891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170135.hg.1 |
ENST00000589437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900170135.hg.1 |
NM_134444.4 |
Refseq |