Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014994.hg.1 |
TC04001089.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014994.hg.1 |
ENST00000282970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014994.hg.1 |
ENST00000382114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014994.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014996.hg.1 |
TC04001089.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014996.hg.1 |
ENST00000282970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014996.hg.1 |
ENST00000382114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014996.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014998.hg.1 |
TC04001089.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014998.hg.1 |
ENST00000282970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014998.hg.1 |
ENST00000382114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014998.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014997.hg.1 |
TC04001089.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014997.hg.1 |
ENST00000382114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014997.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170229.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170229.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170229.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170229.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170229.hg.1 |
TC0400010258.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400170232.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170232.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170232.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170232.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170232.hg.1 |
TC0400010258.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400170231.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170231.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170231.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170231.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170231.hg.1 |
TC0400010258.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000382114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
ENST00000512108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010258.hg.1 |
NM_018176.3 |
Refseq |