Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025184.3 | NP_079460.2 | 11 - 12 | 1832 | 106 | |
XM_006724562.2 | XP_006724625.1 | 10 - 11 | 1545 | 106 | ||
GenBank mRNA | AK026496.1 | - | 11 - 12 | 1686 | 106 | |
AK291187.1 | - | 11 - 12 | 1973 | 106 | ||
AK292967.1 | - | 11 - 12 | 1816 | 106 | ||
BC031039.1 | - | 11 - 12 | 1827 | 5272398 | 106 | |
CR749363.1 | - | 6 - 7 | 1493 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |