Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001872.4 | NP_001863.3 | 7 - 8 | 770 | 148 | |
XM_017020393.1 | XP_016875882.1 | 7 - 8 | 713 | 148 | ||
GenBank mRNA | AK290829.1 | - | 7 - 8 | 718 | 148 | |
BC007057.1 | - | 7 - 8 | 727 | 3934520 | 148 | |
BT006936.1 | - | 7 - 8 | 703 | 148 | ||
M75106.1 | - | 7 - 8 | 722 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons |