Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017026901.1 | XP_016882390.1 | 9 - 10 | 1663 | 102 | |
XM_017026906.1 | XP_016882395.1 | 9 - 10 | 1699 | 102 | ||
GenBank mRNA | AK296781.1 | - | 6 - 7 | 1146 | 102 | |
AL390164.1 | - | 1 - 2 | 111 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |